missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals 2-Methoxy-3-pyridinecarboxaldehyde, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 439780010
Specifications
2-Methoxy-3-pyridinecarboxaldehyde | |
95.0 | |
Colorless to Yellow | |
200°C to 201°C | |
95% min. (GC) | |
C7H7NO2 | |
1g | |
PIFFMIDNNWOQLK-UHFFFAOYSA-N | |
2-methoxypyridine-3-carbaldehyde | |
12533391 | |
Liquid or Low Melting Solid |
71255-09-9 | |
100.0 | |
25°C | |
96°C | |
Authentic | |
Glass bottle | |
2-methoxynicotinaldehyde, 2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-pyridine-3-carbaldehyde, 2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-methoxy, 2-methoxy-3-pyridine carboxaldehyde, 3-pyridinecarboxaldehyde,2-methoxy, 2-methoxy-3-pyridinecarbaldehyde, pubchem15124, 2-methoxy-3-formyl-pyridine | |
COC1=C(C=CC=N1)C=O | |
137.14 | |
137.14 | |
96% |
Chemical Identifiers
71255-09-9 | |
137.14 | |
2-methoxynicotinaldehyde, 2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-pyridine-3-carbaldehyde, 2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-methoxy, 2-methoxy-3-pyridine carboxaldehyde, 3-pyridinecarboxaldehyde,2-methoxy, 2-methoxy-3-pyridinecarbaldehyde, pubchem15124, 2-methoxy-3-formyl-pyridine | |
2-methoxypyridine-3-carbaldehyde |
C7H7NO2 | |
PIFFMIDNNWOQLK-UHFFFAOYSA-N | |
12533391 | |
COC1=C(C=CC=N1)C=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only