Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-3-nitropyridine, 98%

$30.45 - $345.05

Chemical Identifiers

Chemical Name or Material 2-Methoxy-3-nitropyridine
CAS 20265-35-4
Molecular Formula C6H6N2O3
Formula Weight 154.13
MDL Number MFCD00023459
Beilstein 138199
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1830614

  SDS
alfa aesar™
A1830614
25g Each for $345.05

AAA1830603

  SDS
alfa aesar™
A1830603
1g Each for $30.45
Description & Specifications

Specifications

Formula Weight 154.13
MDL Number MFCD00023459
Beilstein 138199
Melting Point 53° to 55°C
Assay 98%
CAS 20265-35-4
Molecular Formula C6H6N2O3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Chemical Name or Material 2-Methoxy-3-nitropyridine
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.