missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54933MD

 View more versions of this product

Catalog No. AAH54933MD



Specifications

Specifications

2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester
250mg
C12H17BN2O5
2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)[N+](=O)[O-]
280.087
280.08
118°C to 119°C
1083168-94-8
MFCD12923397
XWXOPFXUTOQOSM-UHFFFAOYSA-N
2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
57416495
96%

Chemical Identifiers

1083168-94-8
280.087
XWXOPFXUTOQOSM-UHFFFAOYSA-N
57416495
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)[N+](=O)[O-]
C12H17BN2O5
MFCD12923397
2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine
2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents