missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5493303

 View more versions of this product

Catalog No. AAH5493303



Specifications

Specifications

2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester
1g
C12H17BN2O5
2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)[N+](=O)[O-]
280.087
280.08
118°C to 119°C
1083168-94-8
MFCD12923397
XWXOPFXUTOQOSM-UHFFFAOYSA-N
2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
57416495
96%

Chemical Identifiers

1083168-94-8
280.087
XWXOPFXUTOQOSM-UHFFFAOYSA-N
57416495
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)[N+](=O)[O-]
C12H17BN2O5
MFCD12923397
2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine
2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents