missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methoxy-3-methylpyrazine 98.0+%, TCI America™
Supplier: TCI America M081025G
Specifications
2-Methoxy-3-methylpyrazine | |
Yellow | |
C6H8N2O | |
25 g | |
pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a | |
CC1=NC=CN=C1OC | |
124.143 | |
124.14 | |
Liquid |
2847-30-5 | |
87°C | |
MFCD00006127 | |
1993 | |
VKJIAEQRKBQLLA-UHFFFAOYSA-N | |
2-methoxy-3-methylpyrazine | |
17898 | |
≥98.0% (GC) |
Chemical Identifiers
2847-30-5 | |
124.143 | |
VKJIAEQRKBQLLA-UHFFFAOYSA-N | |
17898 | |
CC1=NC=CN=C1OC |
C6H8N2O | |
MFCD00006127 | |
pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a | |
2-methoxy-3-methylpyrazine |
Safety and Handling
TSCA : Yes