Please login to your online account to display your discounted pricing

2-Methoxy-1-naphthonitrile, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  166730050

 View more versions of this product

Catalog No. AC166730050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 183.21
Quantity 5g
Color White to brown
EINECS Number 240-133-3
TSCA TSCA
Melting Point 94° to 98°C
Beilstein 10, II, 205
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 1-Cyano-2-methoxynaphtalene
CAS Min % 97.5
Health Hazard 2 GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methoxy-1-naphthonitrile
Name Note 98%
Assay Percent Range 98%
Molecular Formula C12H9NO
Linear Formula CH3OC10H6CN
MDL Number MFCD00003867
CAS 16000-39-8
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 2-Methoxy-1-naphthonitrile
Name Note 98%
CAS 16000-39-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H9NO
Linear Formula CH3OC10H6CN
Formula Weight 183.21
MDL Number MFCD00003867
Synonym 1-Cyano-2-methoxynaphtalene
Accessories

GHS Signal Word: Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

SDS & Safety Documents