Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-1-naphthoic acid, 98%

Chemical Identifiers

Chemical Name or Material 2-Methoxy-1-naphthoic acid
CAS 947-62-6
Formula Weight 202.21
MDL Number MFCD00021532
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6043403

 
alfa aesar™
H6043403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 177° to 180°C
Formula Weight 202.21
Physical Form Solid
Assay 98%
MDL Number MFCD00021532
Chemical Name or Material 2-Methoxy-1-naphthoic acid
Quantity 1g
CAS 947-62-6