missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-1-naphthalenemethanol, TCI America™

$135.97 - $406.39

Chemical Identifiers

CAS 40696-22-8
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00274224
InChI Key VBHARLNUEDIKQD-UHFFFAOYSA-N
Synonym 2-methoxy-1-naphthalenemethanol, 2-methoxynaphthalen-1-yl methanol, 1-naphthalenemethanol, 2-methoxy, 1-hydroxymethyl-2-methoxynaphthalene, acmc-1ald1, 2-methoxynaphth-1-yl methanol, 2-methoxynaphthyl methan-1-ol, 1-naphthalenemethanol,2-methoxy
PubChem CID 2733640
IUPAC Name (2-methoxynaphthalen-1-yl)methanol
SMILES COC1=C(C2=CC=CC=C2C=C1)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20185G View Documents TCI America
M20185G
5g Each for $135.97
 
M201825G View Documents TCI America
M201825G
25g Each for $406.39
 
Specifications

Chemical Identifiers

40696-22-8
188.226
VBHARLNUEDIKQD-UHFFFAOYSA-N
2733640
COC1=C(C2=CC=CC=C2C=C1)CO
C12H12O2
MFCD00274224
2-methoxy-1-naphthalenemethanol, 2-methoxynaphthalen-1-yl methanol, 1-naphthalenemethanol, 2-methoxy, 1-hydroxymethyl-2-methoxynaphthalene, acmc-1ald1, 2-methoxynaphth-1-yl methanol, 2-methoxynaphthyl methan-1-ol, 1-naphthalenemethanol,2-methoxy
(2-methoxynaphthalen-1-yl)methanol

Specifications

Undesignated
5g
C12H12O2
2-methoxy-1-naphthalenemethanol, 2-methoxynaphthalen-1-yl methanol, 1-naphthalenemethanol, 2-methoxy, 1-hydroxymethyl-2-methoxynaphthalene, acmc-1ald1, 2-methoxynaphth-1-yl methanol, 2-methoxynaphthyl methan-1-ol, 1-naphthalenemethanol,2-methoxy
COC1=C(C2=CC=CC=C2C=C1)CO
188.226
188.23
≥98.0% (GC)
103°C
40696-22-8
MFCD00274224
VBHARLNUEDIKQD-UHFFFAOYSA-N
(2-methoxynaphthalen-1-yl)methanol
2733640
Crystal-Powder at 20°C
2-Methoxy-1-naphthalenemethanol
Safety and Handling

Safety and Handling

TSCA : No