missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-1-naphthalenemethanol 98.0+%, TCI America™

Manufacturer:  Tci America M201825G

 View more versions of this product

Catalog No. M201825G



Specifications

Specifications

2-Methoxy-1-naphthalenemethanol
103°C
40696-22-8
MFCD00274224
VBHARLNUEDIKQD-UHFFFAOYSA-N
(2-methoxynaphthalen-1-yl)methanol
2733640
Crystal-Powder at 20°C
Undesignated
25g
C12H12O2
1-hydroxymethyl-2-methoxynaphthalene, 1-naphthalenemethanol, 2-methoxy, 1-naphthalenemethanol,2-methoxy, 2-methoxy-1-naphthalenemethanol, 2-methoxynaphth-1-yl methanol, 2-methoxynaphthalen-1-yl methanol, 2-methoxynaphthyl methan-1-ol, acmc-1ald1
COC1=C(C2=CC=CC=C2C=C1)CO
188.226
188.23
≥98.0% (GC)

Chemical Identifiers

40696-22-8
188.226
VBHARLNUEDIKQD-UHFFFAOYSA-N
2733640
COC1=C(C2=CC=CC=C2C=C1)CO
C12H12O2
MFCD00274224
1-hydroxymethyl-2-methoxynaphthalene, 1-naphthalenemethanol, 2-methoxy, 1-naphthalenemethanol,2-methoxy, 2-methoxy-1-naphthalenemethanol, 2-methoxynaphth-1-yl methanol, 2-methoxynaphthalen-1-yl methanol, 2-methoxynaphthyl methan-1-ol, acmc-1ald1
(2-methoxynaphthalen-1-yl)methanol
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions