missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1352322

 View more versions of this product

Catalog No. AAL1352322



Description

Description

2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

2-(Methacryloyloxy)ethyltrimethylammonium chloride
5039-78-1
1.105
C9H18ClNO2
MFCD00060097
8168395
It is soluble in water.
CC(=C)C(=O)OCC[N+](C)(C)C.[Cl-]
207.698
207.7
stabilized with 150 to 200ppm 4-methoxyphenol
-25°C
>100°C
1.466
100 g
polyquaternium-37, 2-methacryloyloxy-n,n,n-trimethylethanaminium chloride, unii-pp88r88k3o, methacrylatoethyl trimethyl ammonium chloride, 2-methacryloyloxy ethyl trimethylammonium chloride, 2-methacryloyloxy ethyl trimethylammonium, methacryloxyethyltrimethyl ammonium chloride, ethanaminium, n,n,n-trimethyl-2-2-methyl-1-oxo-2-propenyl oxy-, chloride
RRHXZLALVWBDKH-UHFFFAOYSA-M
trimethyl-[2-(2-methylprop-2-enoyloxy)ethyl]azanium;chloride
78738
72% aq. soln.

Chemical Identifiers

5039-78-1
207.698
RRHXZLALVWBDKH-UHFFFAOYSA-M
78738
CC(=C)C(=O)OCC[N+](C)(C)C.[Cl-]
C9H18ClNO2
MFCD00060097
polyquaternium-37, 2-methacryloyloxy-n,n,n-trimethylethanaminium chloride, unii-pp88r88k3o, methacrylatoethyl trimethyl ammonium chloride, 2-methacryloyloxy ethyl trimethylammonium chloride, 2-methacryloyloxy ethyl trimethylammonium, methacryloxyethyltrimethyl ammonium chloride, ethanaminium, n,n,n-trimethyl-2-2-methyl-1-oxo-2-propenyl oxy-, chloride
trimethyl-[2-(2-methylprop-2-enoyloxy)ethyl]azanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-733-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only