Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methacryloyloxy)ethyl 3,5-dinitrobenzoate, 97%

$134.28 - $134.28

Chemical Identifiers

Chemical Name or Material 2-(Methacryloyloxy)ethyl 3,5-dinitrobenzoate
CAS 78198-10-4
Molecular Formula C13H12N2O8
Formula Weight 324.25g/mol
MDL Number MFCD12546190
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3074514

 
alfa aesar™
H30745-14
25g Each for $134.28
Description & Specifications

Specifications

Melting Point 71° to 72°C
Quantity 25g
Chemical Name or Material 2-(Methacryloyloxy)ethyl 3,5-dinitrobenzoate
CAS 78198-10-4
Assay 97%
Molecular Formula C13H12N2O8
Formula Weight 324.25g/mol
MDL Number MFCD12546190
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.