Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methacryloyloxy)ethyl 3,5-diaminobenzoate, 99%

$128.84 - $463.32

Chemical Identifiers

Chemical Name or Material 2-(Methacryloyloxy)ethyl 3,5-diaminobenzoate
CAS 76067-81-7
Molecular Formula C13H16N2O4
Formula Weight 264.28
MDL Number MFCD00425081
Synonym 3,5-Diaminobenzoic acid 2-(methacryloyloxy)ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2584006

 
alfa aesar™
H2584006
5g Each for $463.32

AAH2584003

 
alfa aesar™
H2584003
1g Each for $128.84
Description & Specifications

Specifications

Formula Weight 264.28
Chemical Name or Material 2-(Methacryloyloxy)ethyl 3,5-diaminobenzoate
Assay 99%
Molecular Formula C13H16N2O4
MDL Number MFCD00425081
CAS 76067-81-7
Synonym 3,5-Diaminobenzoic acid 2-(methacryloyloxy)ethyl ester
Health Hazard 1 Warning
Melting Point 89° to 93°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C