missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mesitylethanol, TCI America™

$44.31 - $143.61

Chemical Identifiers

CAS 6950-92-1
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00020609
InChI Key FQZPTDPHVFTOSY-UHFFFAOYSA-N
Synonym 2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol
PubChem CID 81389
IUPAC Name 2-(2,4,6-trimethylphenyl)ethanol
SMILES CC1=CC(=C(C(=C1)C)CCO)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26245G View Documents TCI America
M26245G
5g Each for $143.61
 
M26241G View Documents TCI America
M26241G
1g Each for $44.31
 
Specifications

Chemical Identifiers

6950-92-1
164.248
FQZPTDPHVFTOSY-UHFFFAOYSA-N
81389
CC1=CC(=C(C(=C1)C)CCO)C
C11H16O
MFCD00020609
2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol
2-(2,4,6-trimethylphenyl)ethanol

Specifications

152°C
80°C
6950-92-1
MFCD00020609
FQZPTDPHVFTOSY-UHFFFAOYSA-N
2-(2,4,6-trimethylphenyl)ethanol
81389
Crystal-Powder at 20°C
2-Mesitylethanol
White
5g
C11H16O
2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol
CC1=CC(=C(C(=C1)C)CCO)C
164.248
164.25
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No