Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mesitylethanol, 98%

$39.03 - $507.78

Chemical Identifiers

Chemical Name or Material 2-Mesitylethanol
CAS 6950-92-1
Molecular Formula C11H16O
Formula Weight 164.25
MDL Number MFCD00020609
Beilstein 1938400
Synonym 2-Hydroxyethylmesitylene; 2,4,6-Trimethylphenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0803203

  SDS
alfa aesar™
L0803203
1g Each for $39.03

AAL0803214

  SDS
alfa aesar™
L0803214
25g Each for $507.78
Description & Specifications

Specifications

Formula Weight 164.25
Chemical Name or Material 2-Mesitylethanol
Assay 98%
CAS 6950-92-1
Molecular Formula C11H16O
MDL Number MFCD00020609
Beilstein 1938400
Quantity 1g
Melting Point 80° to 84°C
Synonym 2-Hydroxyethylmesitylene; 2,4,6-Trimethylphenethyl alcohol