missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mesitylenesulfonyl Hydrazide 97.0+%, TCI America™
$149.32 - $504.33
Chemical Identifiers
CAS | 16182-15-3 |
---|---|
Molecular Formula | C9H14N2O2S |
Molecular Weight (g/mol) | 214.283 |
MDL Number | MFCD00007585 |
InChI Key | JQUBKTQDNVZHIY-UHFFFAOYSA-N |
Synonym | 2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide |
PubChem CID | 85321 |
IUPAC Name | 2,4,6-trimethylbenzenesulfonohydrazide |
SMILES | CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C |
Chemical Identifiers
16182-15-3 | |
214.283 | |
JQUBKTQDNVZHIY-UHFFFAOYSA-N | |
85321 | |
CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C |
C9H14N2O2S | |
MFCD00007585 | |
2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide | |
2,4,6-trimethylbenzenesulfonohydrazide |
Specifications
16182-15-3 | |
White | |
MFCD00007585 | |
1325 | |
JQUBKTQDNVZHIY-UHFFFAOYSA-N | |
2,4,6-trimethylbenzenesulfonohydrazide | |
85321 | |
≥97.0% (T) | |
2-Mesitylenesulfonyl Hydrazide |
114°C | |
C9H14N2O2S | |
5 g | |
2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide | |
CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C | |
214.283 | |
214.28 | |
Crystalline Powder |
Safety and Handling
TSCA : No