Learn More
2-Mesitylenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125595000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Mesitylenesulfonyl chloride | |
773-64-8 | |
100.0 | |
Gray to White | |
>110°C | |
98.5% min. (ex Cl) (Argentometry) | |
C9H11ClO2S | |
MFCD00007434 | |
11, 136 | |
2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride | |
PVJZBZSCGJAWNG-UHFFFAOYSA-N | |
2,4,6-trimethylbenzenesulfonyl chloride | |
13046 | |
99% |
99% | |
98.5 | |
53°C to 57°C | |
150°C (20.0 mmHg) | |
Authentic | |
Plastic bottle | |
(CH3)3C6H2SO2Cl | |
500 g | |
01,661; 05,430; 06,625; 08,318 | |
Solubility in water: reacts. Other solubilities: soluble in ether,toluene and dichloromethane | |
CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C | |
218.7 | |
218.7 | |
Crystalline Powder |
Chemical Identifiers
773-64-8 | |
218.7 | |
PVJZBZSCGJAWNG-UHFFFAOYSA-N | |
13046 | |
CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C |
C9H11ClO2S | |
MFCD00007434 | |
2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride | |
2,4,6-trimethylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 212-257-8
RTECSNumber : DB8930000
TSCA : TSCA
RUO – Research Use Only