Learn More
2-Mercaptothiazoline, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 125565000
Specifications
2-Mercaptothiazoline | |
Plastic bottle | |
103°C to 106°C | |
98% | |
97.5 | |
98% | |
MFCD00126013 | |
2-mercapto-2-thiazoline, 2-mercaptothiazoline, 2-thiazolidenethione, 2-thiazolidinethione, 2-thiazoline-2-thiol, 2-thiothiazolidone, mercaptothiazoline, metabasal, thiazolidine-2-thione, thyroidan | |
WGJCBBASTRWVJL-UHFFFAOYSA-N | |
1,3-thiazolidine-2-thione | |
2723699 | |
Fine Crystalline Powder |
Authentic | |
Beige to White | |
500g | |
96-53-7 | |
100.0 | |
C3H5NS2 | |
27, 140 | |
Solubility in water: soluble in hot water. Other solubilities: soluble in alkaline water and acetic acid,soluble in ethanol,benzene and chcl3,slightly soluble in ether,insoluble in cs2 | |
C1CSC(=S)N1 | |
119.21 | |
119.21 | |
98% |
Chemical Identifiers
96-53-7 | |
119.21 | |
WGJCBBASTRWVJL-UHFFFAOYSA-N | |
2723699 | |
C1CSC(=S)N1 |
C3H5NS2 | |
MFCD00126013 | |
2-mercapto-2-thiazoline, 2-mercaptothiazoline, 2-thiazolidenethione, 2-thiazolidinethione, 2-thiazoline-2-thiol, 2-thiothiazolidone, mercaptothiazoline, metabasal, thiazolidine-2-thione, thyroidan | |
1,3-thiazolidine-2-thione |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-512-1
RTECSNumber : XJ6122000
TSCA : TSCA