missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mercaptopyridine 98.0+%, TCI America™
Supplier: TCI America M024625G
Specifications
2-Mercaptopyridine | |
127°C | |
C5H5NS | |
25 g | |
WHMDPDGBKYUEMW-UHFFFAOYSA-N | |
1H-pyridine-2-thione | |
2723698 | |
111.16 | |
Crystalline Powder |
2637-34-5 | |
Yellow | |
MFCD00006285 | |
2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine | |
C1=CC(=S)NC=C1 | |
111.162 | |
CHEBI:45223 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
2637-34-5 | |
111.162 | |
WHMDPDGBKYUEMW-UHFFFAOYSA-N | |
2723698 | |
1H-pyridine-2-thione |
C5H5NS | |
MFCD00006285 | |
2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine | |
CHEBI:45223 | |
C1=CC(=S)NC=C1 |
Safety and Handling
RTECSNumber : UT9600000
TSCA : Yes