missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptopyridine, TCI America™

$100.83 - $311.67

Chemical Identifiers

CAS 2637-34-5
Molecular Formula C5H5NS
Molecular Weight (g/mol) 111.162
MDL Number MFCD00006285
InChI Key WHMDPDGBKYUEMW-UHFFFAOYSA-N
Synonym 2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
PubChem CID 2723698
ChEBI CHEBI:45223
IUPAC Name 1H-pyridine-2-thione
SMILES C1=CC(=S)NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M024625G View Documents TCI America
M024625G
25g Each for $100.83
 
M0246100G View Documents TCI America
M0246100G
100g Each for $311.67
 
Specifications

Chemical Identifiers

2637-34-5
111.162
WHMDPDGBKYUEMW-UHFFFAOYSA-N
2723698
1H-pyridine-2-thione
C5H5NS
MFCD00006285
2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
CHEBI:45223
C1=CC(=S)NC=C1

Specifications

Yellow
25g
C5H5NS
2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
C1=CC(=S)NC=C1
111.162
CHEBI:45223
Crystal-Powder at 20°C
2-Mercaptopyridine
127°C
2637-34-5
MFCD00006285
WHMDPDGBKYUEMW-UHFFFAOYSA-N
1H-pyridine-2-thione
2723698
111.16
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

RTECSNumber : UT9600000

TSCA : Yes