missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptopyridine N-Oxide Zinc Salt 98.0+%, TCI America™

Manufacturer:  TCI America M0633100G

 View more versions of this product

Catalog No. M0633100G



Specifications

Specifications

2-Mercaptopyridine N-Oxide Zinc Salt
100g
C5H5NOS
2811
317.69
≥98.0% (T)
White-Yellow
13463-41-7
MFCD00067336
Bis(2-pyridylthio) Zinc 1,1′-Dioxide, 1-Hydroxypyridine-2-thione Zinc Salt, 2-Pyridinethiol 1-Oxide Zinc Salt, Zinc Pyrithione
Crystal-Powder at 20°C

Chemical Identifiers

13463-41-7
C5H5NOS
MFCD00067336
Bis(2-pyridylthio) Zinc 1,1′-Dioxide, 1-Hydroxypyridine-2-thione Zinc Salt, 2-Pyridinethiol 1-Oxide Zinc Salt, Zinc Pyrithione
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3725&(9)-1110

RTECSNumber : ZH0950000

TSCA : Yes

Documents