missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 257740010
Specifications
2-Mercaptopyridine-N-oxide, sodium salt | |
Glass bottle | |
109.0°C | |
-30.0°C | |
8.5 to 10.5 (10% soln. at 20°C) | |
3811-73-2,7732-18-5 | |
42.0,60.0 | |
MFCD01941547 | |
2-mercaptopyridine n-oxide sodium salt, 2-pyridinethiol-1-oxide sodium salt, omadine sodium, pyrithione sodium salt, sodium 2-pyridylthio-n-oxide, sodium omadine, sodium omadine van, sodium pyrithione, sodium-2-pyridinethiol-1-oxide, thione reagent | |
XNRNJIIJLOFJEK-UHFFFAOYSA-N | |
sodium;1-oxidopyridine-2-thione | |
19658 | |
Solution |
1.2200g/mL | |
1.22 | |
Yellow to Amber | |
1L | |
40 w/w% Aqueous Solution | |
40.0,58.0 | |
C5H4NNaOS | |
10, 7892 | |
Solubility in water: soluble | |
C1=CC(=S)N(C=C1)[O-].[Na+] | |
149.15 | |
149.15 | |
40 to 42% |
Chemical Identifiers
3811-73-2 | |
149.15 | |
XNRNJIIJLOFJEK-UHFFFAOYSA-N | |
19658 | |
C1=CC(=S)N(C=C1)[O-].[Na+] |
C5H4NNaOS | |
MFCD01941547 | |
2-mercaptopyridine n-oxide sodium salt, 2-pyridinethiol-1-oxide sodium salt, omadine sodium, pyrithione sodium salt, sodium 2-pyridylthio-n-oxide, sodium omadine, sodium omadine van, sodium pyrithione, sodium-2-pyridinethiol-1-oxide, thione reagent | |
sodium;1-oxidopyridine-2-thione |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 223-296-5