missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptopyridine-N-oxide, sodium salt, 40 w/w % aqueous solution, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC257740050

 View more versions of this product

Catalog No. AC257740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Mercaptopyridine-N-oxide, sodium salt
Plastic drum
109.0°C
-30.0°C
8.5 to 10.5 (10% soln. at 20°C)
3811-73-2,7732-18-5
42.0,60.0
MFCD01941547
2-mercaptopyridine n-oxide sodium salt, 2-pyridinethiol-1-oxide sodium salt, omadine sodium, pyrithione sodium salt, sodium 2-pyridylthio-n-oxide, sodium omadine, sodium omadine van, sodium pyrithione, sodium-2-pyridinethiol-1-oxide, thione reagent
XNRNJIIJLOFJEK-UHFFFAOYSA-N
sodium;1-oxidopyridine-2-thione
19658
Solution
1.2200g/mL
1.22
Yellow to Amber
5L
40 w/w% Aqueous Solution
40.0,58.0
C5H4NNaOS
10, 7892
Solubility in water: soluble
C1=CC(=S)N(C=C1)[O-].[Na+]
149.15
149.15
40 to 42%

Chemical Identifiers

3811-73-2
149.15
XNRNJIIJLOFJEK-UHFFFAOYSA-N
19658
C1=CC(=S)N(C=C1)[O-].[Na+]
C5H4NNaOS
MFCD01941547
2-mercaptopyridine n-oxide sodium salt, 2-pyridinethiol-1-oxide sodium salt, omadine sodium, pyrithione sodium salt, sodium 2-pyridylthio-n-oxide, sodium omadine, sodium omadine van, sodium pyrithione, sodium-2-pyridinethiol-1-oxide, thione reagent
sodium;1-oxidopyridine-2-thione
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen

GHS Signal Word: Warning

EINECSNumber : 223-296-5

SDS Documents