missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercaptopyridine, 98%

Manufacturer:  Alfa Aesar™ A1174114

 View more versions of this product

Catalog No. AAA1174114



Description

Description

Applications
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

Solubility
Slightly soluble in chloroform, methanol and dimethyl sulfoxide.

Notes
Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

2-Mercaptopyridine
25g
C5H5NS
105758
WHMDPDGBKYUEMW-UHFFFAOYSA-N
1H-pyridine-2-thione
2723698
111.17
124°C to 130°C
2637-34-5
MFCD00006285
2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
C1=CC(=S)NC=C1
111.162
CHEBI:45223
98%

Chemical Identifiers

2637-34-5
111.162
WHMDPDGBKYUEMW-UHFFFAOYSA-N
2723698
1H-pyridine-2-thione
C5H5NS
MFCD00006285
2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
CHEBI:45223
C1=CC(=S)NC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UT9600000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents