Please login to your online account to display your discounted pricing

2-Mercaptopropionic Acid 95%, ACROS Organics™

$176.11 - $176.11

Chemical Identifiers

Chemical Name or Material 2-Mercaptopropionic acid
Name Note 95%
CAS 79-42-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C3H6O2S
Linear Formula CH3CH(SH)CO2H
Formula Weight 106.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC138790025

  SDS
thermo fisher scientific
AC138790025
Glass bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC138791000

  SDS
thermo fisher scientific
138791000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC138795000

  SDS
thermo fisher scientific
138795000
Glass bottle 500g Each for $176.11
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 106.14
Quantity 2.5kg
Color Clear colorless to slightly yellow
TSCA TSCA
Density 1.1900g/mL
EINECS Number 201-206-5
RTECS Number UF5250000
Beilstein 03, 289
Fieser 15,307
Merck Index 15, 9493
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Mercaptopropionic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%
Refractive Index 1.4804 to 1.4824 (20°C, 589nm) (typically) exact value on the certificate of an
Assay Percent Range 95%
Molecular Formula C3H6O2S
Linear Formula CH3CH(SH)CO2H
MDL Number MFCD00004862
CAS 79-42-5
Synonym Thiolactic acid
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.