Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercaptonicotinic acid, 90+%

$68.42 - $192.86

Chemical Identifiers

Chemical Name or Material 2-Mercaptonicotinic acid
CAS 38521-46-9
Molecular Formula C6H5NO2S
Formula Weight 155.18
MDL Number MFCD00010102
Beilstein 119029
Synonym 2-Mercaptopyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1468314

  SDS
alfa aesar™
A1468314
25g Each for $68.42

AAA1468322

  SDS
alfa aesar™
A1468322
100g Each for $192.86
Description & Specifications

Specifications

Formula Weight 155.18
TSCA TSCA
MDL Number MFCD00010102
Synonym 2-Mercaptopyridine-3-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Melting Point ≈270°C (dec.)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥90%
Chemical Name or Material 2-Mercaptonicotinic acid
Molecular Formula C6H5NO2S
CAS 38521-46-9
Beilstein 119029
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.