missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mercaptoethyl sulfide, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 125490100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Mercaptoethyl sulfide | |
1.1830g/mL | |
90°C | |
88 % min. (GC) | |
C4H10S3 | |
(HSCH2CH2)2S | |
10g | |
2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi | |
KSJBMDCFYZKAFH-UHFFFAOYSA-N | |
2-(2-sulfanylethylsulfanyl)ethanethiol | |
77117 | |
90% |
3570-55-6 | |
135.0°C to 136.0°C (10.0mmHg) | |
Authentic | |
Glass bottle | |
1.594 | |
MFCD00004893 | |
1.183 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,toluene,chloroform,,dioxane,carbon tetrachloride | |
SCCSCCS | |
154.30 | |
154.32 |
Chemical Identifiers
3570-55-6 | |
154.30 | |
KSJBMDCFYZKAFH-UHFFFAOYSA-N | |
77117 | |
SCCSCCS |
C4H10S3 | |
MFCD00004893 | |
2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi | |
2-(2-sulfanylethylsulfanyl)ethanethiol |
Safety and Handling
Warning
EINECSNumber : 222-671-
RUO â Research Use Only