Please login to your online account to display your discounted pricing

2-Mercaptoethanol, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$104.08 - $104.08

Chemical Identifiers

Chemical Name or Material 2-Mercaptoethanol
CAS 60-24-2
Purity Grade Ultrapure Bioreagent
Molecular Formula HSCH2CH2OH
Formula Weight 78.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002078

 
avantor performance
404900
Steel Can 100g Each for $104.08
Description & Specifications

Specifications

Formula Weight 78.13
Chemical Name or Material 2-Mercaptoethanol
Specific Gravity 1L = 1.11kg
Molecular Formula HSCH2CH2OH
CAS 60-24-2
Quantity 100g
Flash Point 74.00°C
Packaging Steel Can
Purity Grade Ultrapure Bioreagent

    For Electrophoresis and other Molecular Biology Applications