Please login to your online account to display your discounted pricing

2-Mercaptoethanol, MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  806444

 View more versions of this product

Catalog No. ICN806444

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Synonym 2-Hydroxy-1-ethanethiol
CAS 60-24-2
Chemical Name or Material 2-Mercaptoethanol, 25g

Specifications

Chemical Name or Material 2-Mercaptoethanol, 25g
CAS 60-24-2
Synonym 2-Hydroxy-1-ethanethiol