Please login to your online account to display your discounted pricing

2-Mercaptoethanol, MP Biomedicals


Quantity 25g

Manufacturer: mp biomedicals  04806443

 View more versions of this product

Catalog No. ICN806443

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 25g
Chemical Name or Material 2-Mercaptoethanol, 25g
CAS 60-24-2
Synonym 2-Hydroxy-1-ethanethiol

Specifications

Chemical Name or Material 2-Mercaptoethanol, 25g
CAS 60-24-2
Synonym 2-Hydroxy-1-ethanethiol