Please login to your online account to display your discounted pricing

2-Mercaptoethanol, MP Biomedicals

$80.85 - $80.85

Chemical Identifiers

Chemical Name or Material 2-Mercaptoethanol, 25g
CAS 60-24-2
Synonym 2-Hydroxy-1-ethanethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN806444

 
mp biomedicals
806444
100g Each for $80.85
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material 2-Mercaptoethanol, 25g
CAS 60-24-2
Synonym 2-Hydroxy-1-ethanethiol