missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptoethanol (Electrophoresis), Fisher BioReagents

Manufacturer:  Fisher BioReagents FLBP176100

Catalog No. BP176-100



Description

Description

2-Mercaptoethanol reduces disulfide linkages in proteins and peptides and is used in many enzymatic reactions as a protein stabilizer.
Specifications

Specifications

2-Mercaptoethanol
60-24-2
1.110g/cm³
Yellow
157°C
≥98 %
HSCH2CH2OH
Amber Glass/Poison Pack
89.74mL
DGVVWUTYPXICAM-UHFFFAOYSA-N
2-sulfanylethanol
1567
78.13
≥98%
2.69g/mol
Mercaptoethanol
0.05 AU max.
5.2
-100°C
Pass Test
C2H6OS
MFCD00004890
15, 5936
2-hydroxy-1-ethanethiol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan, 2-mercaptoethanol, 2-thioethanol, beta-mercaptoethanol, ethanol, 2-mercapto, mercaptoethanol, thioethylene glycol, thioglycol
OCCS
78.13
CHEBI:41218
Liquid
Electrophoresis

Chemical Identifiers

60-24-2
78.13
DGVVWUTYPXICAM-UHFFFAOYSA-N
1567
2-sulfanylethanol
C2H6OS
MFCD00004890
2-hydroxy-1-ethanethiol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan, 2-mercaptoethanol, 2-thioethanol, beta-mercaptoethanol, ethanol, 2-mercapto, mercaptoethanol, thioethylene glycol, thioglycol
CHEBI:41218
OCCS
Safety and Handling

Safety and Handling

Recommended Storage : Refrigeration

SDS Documents
Product Certifications
Promotions

Promotions