Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercaptobenzothiazole, Ultrapure MALDI Matrix

For use as a matrix in MALDI mass spectrometry analysis

Chemical Identifiers

Chemical Name or Material 2-Mercaptobenzothiazole
CAS 149-30-4
Name Note Ultrapure MALDI Matrix
Formula Weight 167.25
MDL Number MFCD00005781
UN Number UN3077
Beilstein 119484
Merck Index 14,5868
Synonym 2-Mercaptobenzothiazole / Benzothiazole-2-thiol; MBT
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67540EXK

  SDS
alfa aesar™
J67540EXK
5 x 10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 178° to 182°C
Formula Weight 167.25
Chemical Name or Material 2-Mercaptobenzothiazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Physical Form Powder
UN Number UN3077
MDL Number MFCD00005781
Color Pale yellow to cream
RTECS Number DL6475000
Name Note Ultrapure MALDI Matrix
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Synonym 2-Mercaptobenzothiazole / Benzothiazole-2-thiol; MBT
Merck Index 14,5868
Sensitivity Air Sensitive
Beilstein 119484
TSCA TSCA
Density 1.420g/mL
Health Hazard 3 GHS P Statement
P261-P280-P301+P312-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5 x 10mg
CAS 149-30-4
Safety and Handling

Warning

GHS P Statement
P261-P280-P301+P312-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.