missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mercaptobenzothiazole 98.0+%, TCI America™
Supplier: TCI America M0055500G
Specifications
2-Mercaptobenzothiazole | |
180°C | |
C7H5NS2 | |
500 g | |
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
C1=CC=C2C(=C1)NC(=S)S2 | |
167.244 | |
CHEBI:34292 | |
≥98.0% (HPLC,T) |
149-30-4 | |
White-Yellow | |
MFCD00005781 | |
3077 | |
YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
3H-1,3-benzothiazole-2-thione | |
697993 | |
167.24 | |
Crystalline Powder |
Chemical Identifiers
149-30-4 | |
167.244 | |
YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
697993 | |
3H-1,3-benzothiazole-2-thione |
C7H5NS2 | |
MFCD00005781 | |
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
CHEBI:34292 | |
C1=CC=C2C(=C1)NC(=S)S2 |
Safety and Handling
EINECSNumber : (5)-0242
RTECSNumber : DL6475000
TSCA : Yes