missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptobenzothiazole, TCI America™

Manufacturer:  TCI America M024725G

 View more versions of this product

Catalog No. M024725G



Specifications

Specifications

2-Mercaptobenzothiazole
180°C
149-30-4
MFCD00005781
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
C1=CC=C2C(=C1)NC(=S)S2
167.244
CHEBI:34292
Crystal-Powder at 20°C
White-Yellow
25g
C7H5NS2
3077
YXIWHUQXZSMYRE-UHFFFAOYSA-N
3H-1,3-benzothiazole-2-thione
697993
167.24
≥99.0% (HPLC)

Chemical Identifiers

149-30-4
167.244
YXIWHUQXZSMYRE-UHFFFAOYSA-N
697993
3H-1,3-benzothiazole-2-thione
C7H5NS2
MFCD00005781
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
CHEBI:34292
C1=CC=C2C(=C1)NC(=S)S2
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0242

RTECSNumber : DL6475000

TSCA : Yes

Documents