Learn More
2-Mercaptobenzothiazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 332050010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
2-Mercaptobenzothiazole | |
149-30-4 | |
100.0 | |
Yellow | |
Authentic | |
Plastic bottle | |
MFCD00005781 | |
27, II, 233 | |
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
3H-1,3-benzothiazole-2-thione | |
697993 | |
167.24 | |
Powder |
98% | |
97.5 | |
172.0°C to 182.0°C | |
243°C | |
97% | |
C7H5NS2 | |
1 kg | |
15, 5935 | |
yellow solution | |
C1=CC=C2C(=C1)NC(=S)S2 | |
167.24 | |
CHEBI:34292 | |
97% |
Chemical Identifiers
149-30-4 | |
167.24 | |
YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
697993 | |
3H-1,3-benzothiazole-2-thione |
C7H5NS2 | |
MFCD00005781 | |
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
CHEBI:34292 | |
C1=CC=C2C(=C1)NC(=S)S2 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves.
GHS Signal Word: Warning
EINECSNumber : 205-736-8
RTECSNumber : DL6475000
TSCA : TSCA
RUO – Research Use Only