missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptobenzothiazole, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 125452500

 View more versions of this product

Catalog No. AC125452500



Description

Description

Indicator
Specifications

Specifications

2-Mercaptobenzothiazole
Authentic
Plastic bottle
Yellow
250g
94.0
95%
MFCD00005781
15, 5935
Solubility in water: 0.12g/L (25°C). Other solubilities: soluble in alkalies,soluble in alcohol,acetone,benzene and,chloroform
C1=CC=C2C(=C1)NC(=S)S2
167.24
CHEBI:34292
Powder
243°C
0.5% max. (60°C, 3 hrs)
0.5% max.
172.0°C to 182.0°C
149-30-4
100.0
C7H5NS2
27, II, 233
1,3-benzothiazole-2-thiol, 2 3h-benzothiazolethione, 2-benzothiazolethiol, 2-mercaptobenzothiazole, benzo d thiazole-2-thiol, benzothiazole-2-thiol, benzothiazolethiol, captax, rokon, rotax
YXIWHUQXZSMYRE-UHFFFAOYSA-N
3H-1,3-benzothiazole-2-thione
697993
167.24
95%

Chemical Identifiers

149-30-4
167.24
YXIWHUQXZSMYRE-UHFFFAOYSA-N
697993
3H-1,3-benzothiazole-2-thione
C7H5NS2
MFCD00005781
1,3-benzothiazole-2-thiol, 2 3h-benzothiazolethione, 2-benzothiazolethiol, 2-mercaptobenzothiazole, benzo d thiazole-2-thiol, benzothiazole-2-thiol, benzothiazolethiol, captax, rokon, rotax
CHEBI:34292
C1=CC=C2C(=C1)NC(=S)S2
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves.

GHS Signal Word: Warning

EINECSNumber : 205-736-8

SDS Documents
Product Certifications