missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercaptobenzimidazole, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 125442500

 View more versions of this product

Catalog No. AC125442500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Mercaptobenzimidazole
583-39-1
100.0
Beige to White
>250°C
98%
C7H6N2S
250 g
15, 1084
Solubility in water: 2.6g/L (23°C). Other solubilities: soluble in acetone,ethyl acetate and ethanol
S=C1NC2=CC=CC=C2N1
150.20
150.2
0.3% max. (in NaOH)
98%
97.5
302°C to 307°C
451°C
Authentic
Plastic bottle
MFCD00466107
24, 119
2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb
YHMYGUUIMTVXNW-UHFFFAOYSA-N
1,3-dihydrobenzimidazole-2-thione
707035
98%
Powder

Chemical Identifiers

583-39-1
150.20
YHMYGUUIMTVXNW-UHFFFAOYSA-N
707035
C7H6N2S
MFCD00466107
2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb
S=C1NC2=CC=CC=C2N1
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 209-502-6

RTECSNumber : DE1050000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only