Please login to your online account to display your discounted pricing

2-mercapto-6-propylpyrimidin-4-ol, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Mercapto-6-propylpyrimidin-4-ol
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2OS
Formula Weight 170.24
Synonym 6-Propyl-2-thiouracil
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB00664EE

  SDS
fisher scientific company
SB00664EE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.24
Quantity 50g
Assay 97%
Molecular Formula C7H10N2OS
Chemical Name or Material 2-Mercapto-6-propylpyrimidin-4-ol
Assay Percent Range 97%
CAS 51-52-5
Synonym 6-Propyl-2-thiouracil
Health Hazard 1 Warning
Safety and Handling

Warning