Please login to your online account to display your discounted pricing

2-mercapto-6-propylpyrimidin-4-ol, Maybridge


Quantity 50g

Manufacturer: fisher scientific company  SB00664EE

 View more versions of this product

Catalog No. SB00664EE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 170.24
Quantity 50g
Assay 97%
Molecular Formula C7H10N2OS
Chemical Name or Material 2-Mercapto-6-propylpyrimidin-4-ol
Assay Percent Range 97%
CAS 51-52-5
Synonym 6-Propyl-2-thiouracil
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2-Mercapto-6-propylpyrimidin-4-ol
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2OS
Formula Weight 170.24
Synonym 6-Propyl-2-thiouracil
Accessories

Warning