missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Mercapto-6-nitrobenzothiazole 96%, ACROS Organics™

$289.18 - $289.18

Chemical Identifiers

CAS 4845-58-3
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C7H4N2O2S2
Formula Weight 212.25
MDL Number MFCD00041850
Chemical Name or Material 2-Mercapto-6-nitrobenzothiazole, 96%
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC413610050 SDS Thermo Fisher Scientific
413610050
Glass bottle 5g Each for $289.18
Specifications Description & Specifications

Chemical Identifiers

4845-58-3
100.0
C7H4N2O2S2
MFCD00041850
≥95%
95.0
96%
212.25
2-Mercapto-6-nitrobenzothiazole, 96%

Specifications

4845-58-3
100.0
Glass bottle
96%
212.25
Yellow
5g
≥95%
95.0
Authentic
Powder
C7H4N2O2S2
MFCD00041850
247° to 255°C (dec.)
2-Mercapto-6-nitrobenzothiazole, 96%