Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-6-nitrobenzothiazole, 96%

$83.18 - $83.18

Chemical Identifiers

Chemical Name or Material 2-Mercapto-6-nitrobenzothiazole
CAS 4845-58-3
Molecular Formula C7H4N2O2S2
Formula Weight 212.24
MDL Number MFCD00041850
Synonym 6-Nitrobenzothiazole-2-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1821103

  SDS
alfa aesar™
A1821103
1g Each for $83.18
Description & Specifications

Specifications

Formula Weight 212.24
Chemical Name or Material 2-Mercapto-6-nitrobenzothiazole
TSCA TSCA
Assay 96%
Molecular Formula C7H4N2O2S2
MDL Number MFCD00041850
CAS 4845-58-3
Synonym 6-Nitrobenzothiazole-2-thiol
Health Hazard 1 Warning
Melting Point 250° to 254°C
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DL6600000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.