missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mercapto-5-nitrobenzimidazole 98.0+%, TCI America™
$225.62 - $900.84
Chemical Identifiers
CAS | 6325-91-3 |
---|---|
Molecular Formula | C7H5N3O2S |
Molecular Weight (g/mol) | 195.20 |
MDL Number | MFCD00804727 |
InChI Key | YPXQSGWOGQPLQO-UHFFFAOYSA-N |
Synonym | 2-mercapto-5-nitrobenzimidazole, 5-nitro-2-benzimidazolethiol, 5-nitro-2-mercaptobenzimidazole, 5-nitro-1h-benzo d imidazole-2-thiol, 2-benzimidazolethiol, 5-nitro, 2-benzimidazolinethione, 5-nitro, 2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro, 6-nitro-1h-benzimidazole-2-thiol, 5-nitro-1h-benzimidazole-2-thiol, 5-nitro-3-hydrobenzimidazole-2-thione |
PubChem CID | 3717598 |
IUPAC Name | 5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione |
SMILES | [O-][N+](=O)C1=CC=C2NC(=S)NC2=C1 |
Chemical Identifiers
6325-91-3 | |
195.20 | |
YPXQSGWOGQPLQO-UHFFFAOYSA-N | |
3717598 | |
[O-][N+](=O)C1=CC=C2NC(=S)NC2=C1 |
C7H5N3O2S | |
MFCD00804727 | |
2-mercapto-5-nitrobenzimidazole, 5-nitro-2-benzimidazolethiol, 5-nitro-2-mercaptobenzimidazole, 5-nitro-1h-benzo d imidazole-2-thiol, 2-benzimidazolethiol, 5-nitro, 2-benzimidazolinethione, 5-nitro, 2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro, 6-nitro-1h-benzimidazole-2-thiol, 5-nitro-1h-benzimidazole-2-thiol, 5-nitro-3-hydrobenzimidazole-2-thione | |
5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione |
Specifications
6325-91-3 | |
MFCD00804727 | |
2-mercapto-5-nitrobenzimidazole, 5-nitro-2-benzimidazolethiol, 5-nitro-2-mercaptobenzimidazole, 5-nitro-1h-benzo d imidazole-2-thiol, 2-benzimidazolethiol, 5-nitro, 2-benzimidazolinethione, 5-nitro, 2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro, 6-nitro-1h-benzimidazole-2-thiol, 5-nitro-1h-benzimidazole-2-thiol, 5-nitro-3-hydrobenzimidazole-2-thione | |
[O-][N+](=O)C1=CC=C2NC(=S)NC2=C1 | |
195.20 | |
195.20 | |
Crystalline Powder |
C7H5N3O2S | |
5 g | |
YPXQSGWOGQPLQO-UHFFFAOYSA-N | |
5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione | |
3717598 | |
≥98.0% (HPLC) | |
2-Mercapto-5-nitrobenzimidazole |
Safety and Handling
TSCA : Yes