missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Mercapto-5-nitrobenzimidazole 98.0+%, TCI America™
Supplier: TCI America M132525G
Specifications
2-Mercapto-5-nitrobenzimidazole | |
C7H5N3O2S | |
25 g | |
YPXQSGWOGQPLQO-UHFFFAOYSA-N | |
5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione | |
3717598 | |
≥98.0% (HPLC) |
6325-91-3 | |
MFCD00804727 | |
2-mercapto-5-nitrobenzimidazole, 5-nitro-2-benzimidazolethiol, 5-nitro-2-mercaptobenzimidazole, 5-nitro-1h-benzo d imidazole-2-thiol, 2-benzimidazolethiol, 5-nitro, 2-benzimidazolinethione, 5-nitro, 2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro, 6-nitro-1h-benzimidazole-2-thiol, 5-nitro-1h-benzimidazole-2-thiol, 5-nitro-3-hydrobenzimidazole-2-thione | |
[O-][N+](=O)C1=CC=C2NC(=S)NC2=C1 | |
195.20 | |
195.20 | |
Crystalline Powder |
Chemical Identifiers
6325-91-3 | |
195.20 | |
YPXQSGWOGQPLQO-UHFFFAOYSA-N | |
3717598 | |
[O-][N+](=O)C1=CC=C2NC(=S)NC2=C1 |
C7H5N3O2S | |
MFCD00804727 | |
2-mercapto-5-nitrobenzimidazole, 5-nitro-2-benzimidazolethiol, 5-nitro-2-mercaptobenzimidazole, 5-nitro-1h-benzo d imidazole-2-thiol, 2-benzimidazolethiol, 5-nitro, 2-benzimidazolinethione, 5-nitro, 2h-benzimidazole-2-thione, 1,3-dihydro-5-nitro, 6-nitro-1h-benzimidazole-2-thiol, 5-nitro-1h-benzimidazole-2-thiol, 5-nitro-3-hydrobenzimidazole-2-thione | |
5-nitro-2,3-dihydro-1H-1,3-benzodiazole-2-thione |
Safety and Handling
TSCA : Yes