Please login to your online account to display your discounted pricing

2-Mercapto-5-nitrobenzimidazole, 97%, ACROS Organics™

$29.02 - $29.02

Chemical Identifiers

Chemical Name or Material 2-Mercapto-5-nitrobenzimidazole
Name Note 97%
CAS 6325-91-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H5N3O2S
Formula Weight 195.2
MDL Number MFCD00804727
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334480050

  SDS
thermo fisher scientific
334480050
Glass bottle 5g Each for $29.02
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 195.2
Melting Point 272.0° to 278.0°C (dec.)
Quantity 5g
Color Yellow-brown
TSCA TSCA
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 6325-91-3
Chemical Name or Material 2-Mercapto-5-nitrobenzimidazole
Molecular Formula C7H5N3O2S
Assay Percent Range 97%
Name Note 97%
MDL Number MFCD00804727
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.