Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-5-nitrobenzimidazole, 97%

$126.88 - $438.47

Chemical Identifiers

Chemical Name or Material 2-Mercapto-5-nitrobenzimidazole
CAS 6325-91-3
Molecular Formula C7H5N3O2S
Formula Weight 195.2
MDL Number MFCD00804727
Beilstein 172127
Synonym 5-Nitro-2-benzimidazolethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2235814

  SDS
alfa aesar™
B2235814
25g Each for $126.88

AAB2235822

  SDS
alfa aesar™
B2235822
100g Each for $438.47
Description & Specifications

Specifications

Formula Weight 195.2
Chemical Name or Material 2-Mercapto-5-nitrobenzimidazole
TSCA TSCA
Assay 97%
Molecular Formula C7H5N3O2S
MDL Number MFCD00804727
CAS 6325-91-3
Beilstein 172127
Synonym 5-Nitro-2-benzimidazolethiol
Health Hazard 1 Warning
Melting Point ≈273°C (dec.)
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.