Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-5-methyl-1,3,4-thiadiazole, 99%

$319.43 - $319.43

Chemical Identifiers

Chemical Name or Material 2-Mercapto-5-methyl-1,3,4-thiadiazole
CAS 29490-19-5
Molecular Formula C3H4N2S2
Formula Weight 132.21
MDL Number MFCD00038349
Beilstein 606663
Synonym 5-Methyl-1,3,4-thiadiazole-2-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2135530

  SDS
alfa aesar™
B2135530
250g Each for $319.43
Description & Specifications

Specifications

Formula Weight 132.21
Chemical Name or Material 2-Mercapto-5-methyl-1,3,4-thiadiazole
Assay 99%
Molecular Formula C3H4N2S2
MDL Number MFCD00038349
CAS 29490-19-5
Beilstein 606663
Synonym 5-Methyl-1,3,4-thiadiazole-2-thiol
Health Hazard 1 Warning
Melting Point 182° to 185°C
Quantity 250g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.