Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-5-methoxy-3H-imidazo[4,5-b]pyridine, 95%

$129.27 - $129.27

Chemical Identifiers

Chemical Name or Material 2-Mercapto-5-methoxy-3H-imidazo[4,5-b]pyridine
CAS 113713-60-3
Molecular Formula C7H7N3OS
Formula Weight 181.22
MDL Number MFCD06658230
Synonym 5-Methoxy-2H-imidazo[4,5-b]pyridine-2-thione; 1,3-Dihydro-5-methoxy-2H-imidazo[4,5-b]pyridine-2-thione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2660803

  SDS
alfa aesar™
H2660803
1g Each for $129.27
Description & Specifications

Specifications

Formula Weight 181.22
Health Hazard 1 Warning
Quantity 1g
MDL Number MFCD06658230
Molecular Formula C7H7N3OS
Chemical Name or Material 2-Mercapto-5-methoxy-3H-imidazo[4,5-b]pyridine
Synonym 5-Methoxy-2H-imidazo[4,5-b]pyridine-2-thione; 1,3-Dihydro-5-methoxy-2H-imidazo[4,5-b]pyridine-2-thione
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 238° to 241°C
Assay 95%
CAS 113713-60-3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.