missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-4-(trifluoromethyl)pyrimidine, 96%

$64.00 - $272.96

Chemical Identifiers

CAS 136547-17-6
Molecular Formula C5H3F3N2S
Molecular Weight (g/mol) 180.148
MDL Number MFCD00115113
InChI Key HDGJASCXBNRLGA-UHFFFAOYSA-N
Synonym 4-trifluoromethyl pyrimidine-2-thiol, 4-trifluoromethyl-2-pyrimidinethiol, 2-mercapto-4-trifluoromethylpyrimidine, 2-mercapto-4-trifluoromethyl pyrimidine, 6-trifluoromethyl-1h-pyrimidine-2-thione, 2 1h-pyrimidinethione, 4-trifluoromethyl, 4-trifluoromethyl-3-hydropyrimidine-2-thione, 4-trifluoromethyl-3h-pyrimidine-2-thione, zlchem 646, pubchem17679
PubChem CID 1240105
IUPAC Name 6-(trifluoromethyl)-1H-pyrimidine-2-thione
SMILES C1=C(NC(=S)N=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6050103 View Documents Alfa Aesar™
H6050103
1g Each for $64.00
 
AAH6050106 View Documents Alfa Aesar™
H6050106
5g Each for $272.96
 
Specifications

Chemical Identifiers

136547-17-6
180.148
HDGJASCXBNRLGA-UHFFFAOYSA-N
1240105
C1=C(NC(=S)N=C1)C(F)(F)F
C5H3F3N2S
MFCD00115113
4-trifluoromethyl pyrimidine-2-thiol, 4-trifluoromethyl-2-pyrimidinethiol, 2-mercapto-4-trifluoromethylpyrimidine, 2-mercapto-4-trifluoromethyl pyrimidine, 6-trifluoromethyl-1h-pyrimidine-2-thione, 2 1h-pyrimidinethione, 4-trifluoromethyl, 4-trifluoromethyl-3-hydropyrimidine-2-thione, 4-trifluoromethyl-3h-pyrimidine-2-thione, zlchem 646, pubchem17679
6-(trifluoromethyl)-1H-pyrimidine-2-thione

Specifications

164°C to 166°C
136547-17-6
MFCD00115113
4-trifluoromethyl pyrimidine-2-thiol, 4-trifluoromethyl-2-pyrimidinethiol, 2-mercapto-4-trifluoromethylpyrimidine, 2-mercapto-4-trifluoromethyl pyrimidine, 6-trifluoromethyl-1h-pyrimidine-2-thione, 2 1h-pyrimidinethione, 4-trifluoromethyl, 4-trifluoromethyl-3-hydropyrimidine-2-thione, 4-trifluoromethyl-3h-pyrimidine-2-thione, zlchem 646, pubchem17679
C1=C(NC(=S)N=C1)C(F)(F)F
180.148
180.15
2-Mercapto-4-(trifluoromethyl)pyrimidine
1g
C5H3F3N2S
UN2811
HDGJASCXBNRLGA-UHFFFAOYSA-N
6-(trifluoromethyl)-1H-pyrimidine-2-thione
1240105
96%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.