Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-4-phenylthiazole, 98%

$110.36 - $410.99

Chemical Identifiers

Chemical Name or Material 2-Mercapto-4-phenylthiazole
CAS 2103-88-0
Molecular Formula C9H7NS2
Formula Weight 193.3
MDL Number MFCD00014508
Beilstein 128288
Synonym 4-Phenyl-2-thiazolethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0582906

  SDS
alfa aesar™
L0582906
5g Each for $110.36

AAL0582914

  SDS
alfa aesar™
L0582914
25g Each for $410.99
Description & Specifications

Specifications

Formula Weight 193.3
CAS 2103-88-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Mercapto-4-phenylthiazole
TSCA TSCA
Assay 98%
Molecular Formula C9H7NS2
Synonym 4-Phenyl-2-thiazolethiol
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XJ5123450
MDL Number MFCD00014508
Beilstein 128288
Melting Point 172° to 175°C
Quantity 5g
Odor Odorless
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.