Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Mercapto-4-methylthiazole, 97%

$67.81 - $739.82

Chemical Identifiers

Chemical Name or Material 2-Mercapto-4-methylthiazole
CAS 4498-39-9
Molecular Formula C4H5NS2
Formula Weight 117.2
MDL Number MFCD00798571
Synonym 4-Methylthiazole-2-thiol; 4-Methyl-2(3H)-thiazolethione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1952014

  SDS
alfa aesar™
A1952014
25g Each for $67.81

AAA1952022

  SDS
alfa aesar™
A1952022
100g Each for $213.96

AAA1952036

  SDS
alfa aesar™
A1952036
500g Each for $739.82
Description & Specifications

Specifications

Formula Weight 117.2
MDL Number MFCD00798571
Assay 97%
CAS 4498-39-9
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 86° to 89°C
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C4H5NS2
TSCA TSCA
Health Hazard 1 Warning
Chemical Name or Material 2-Mercapto-4-methylthiazole
Synonym 4-Methylthiazole-2-thiol; 4-Methyl-2(3H)-thiazolethione
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.